missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRA-1-81 (Podocalyxin) Monoclonal Antibody (TRA-1-81), APC, eBioscience™
GREENER_CHOICE

Product Code. 15588206
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15588206 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15588206 Supplier Invitrogen™ Supplier No. 17888342

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: The TRA-1-81 antibody recognizes a protein expressed on undifferentiated human embryonic stem cells (ES), embyronal carcinoma cells (EC), and embryonic germ cells (EG). Like other stem cell specific markers, the epitope recognized by the TRA-1-81 antibody is lost upon cell differentiation. The TRA-1-81 epitope is resistant to neuraminidase digestion, unlike the epitope recognized by the related TRA-1-60 antibody. The TRA-1-81 antibody is known to specifically recognize a carbohydrate epitope on a keratan sulfated glycoprotein recently identified as podocalyxin, a member of the CD34-related family of sialomucins. Podocalyxin is a transmembrane glycoprotein originally identified on epithelial glomerular cells known as podocytes, and the protein has also been implicated in the development of aggressiveness in a variety of cancers, including breast and prostate cancer. Applications Reported: This TRA-1-81 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TRA-1-81 antibody has been pre-titrated and tested by flow cytometric analysis of 2102Ep human embryonal carcinoma cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing...

TRA-1-81 is a cell surface antigen expressed along with SSEA-3, SSEA-4 and TRA-1-60 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are down-regulated during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRA-1-81 (Podocalyxin)
Applications Flow Cytometry
Classification Monoclonal
Clone TRA-1-81
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Alias GCTM-2 antigen; Gp200; PC; PCLP; PCLP1; PCLP-1; Podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; PODXL; TRA180; tra1-81
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.