missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRA-1-81 Mouse, anti-Human, Alexa Fluor 488, Clone: TRA-1-81, STEMCELL Technologies
Mouse Monoclonal Antibody
£283.50 - £561.60
Specifications
| Antigen | TRA-1-81 |
|---|---|
| Clone | TRA-1-81 |
| Applications | Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17198801
|
Stemcell Technologies
60065AD.1 |
25 Tests |
£283.50
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17188801
|
Stemcell Technologies
60065AD |
100 Tests |
£561.60
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The TRA-1-81 antibody reacts with a carbohydrate epitope on TRA-1-81, a high-molecular-mass (> 200 kDa) glycoprotein expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey and rat ES cell lines
- No immunoreactivity is observed with mouse EC, EG, ES, or iPS cells
- The epitope of the TRA-1-81 antibody is associated with a keratan-sulfated transmembrane protein identified as podocalyxin and is resistant to treatment with neuraminidase
Specifications
| TRA-1-81 | |
| Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry | |
| Alexa Fluor 488 | |
| Rhesus Monkey, Human, Rat | |
| Primary |
| TRA-1-81 | |
| Monoclonal | |
| Mouse | |
| IgM κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title