missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRA-1-81 Monoclonal Antibody (TRA-1-81)

Mouse Monoclonal Antibody
£188.00 - £387.00
Specifications
| Antigen | TRA-1-81 |
|---|---|
| Clone | TRA-1-81 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12837752
![]() |
Invitrogen™
MA1024 |
100 μg |
£387.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
12847752
|
Invitrogen™
MA1024X |
20 μL |
£188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRA-1-81 Monoclonal Antibody for Western Blot, ICC/IF, Flow
TRA-1-81 is a cell surface antigen expressed along with SSEA-3, SSEA-4 and TRA-1-60 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are down-regulated during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.Specifications
| TRA-1-81 | |
| 1 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| human embryonal carcinoma cell line 2102Ep. | |
| Antibody |
| TRA-1-81 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Liquid | |
| IgM | |
| Human | |
| GCTM-2 antigen; Gp200; PC; PCLP; PCLP1; PCLP-1; Podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; PODXL; TRA180; tra1-81 | |
| Primary | |
| Affinity chromatography - MBP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title