missing translation for 'onlineSavingsMsg'
Learn More

TRA-1-81 Antibody (TRA-1-81) - Azide and BSA Free, Novus Biologicals™

Product Code. 18335020 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335020 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335020 Supplier Novus Biologicals Supplier No. NBP280986

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRA-1-81 Monoclonal specifically detects TRA-1-81 in Human, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRA-1-81
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TRA-1-81
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:500, Flow Cytometry 1:10-1:1000. Use reported in scientific literature, Immunohistochemistry : 1:50-1:200, Immunocytochemistry/Immunofluorescence : 1:50-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin : 1:50-1:200, Immunohistochemistry-Frozen : 1:50-1:200
Formulation Tris-Glycine, 0.15 M NaCl
Gene Alias gp200, MGC138240, PC, PCLP, PODXL
Host Species Mouse
Immunogen Human embryonal carcinoma cell line 2102Ep.
Purification Method IgM purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Human. This antibody recognizes the TRA-1-81 antigen that is expressed upon the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES). No immunoreactivity is seen with murine EC, EG or ES cells. This antibody recognizes antigens that are associated with a pericellular matrix proteoglycan.
Target Species Human, Mouse (Negative)
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.