missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRA-1-60 Monoclonal Antibody (TRA-1-60)

Description
TRA-1-60 Monoclonal Antibody for Flow, ICC/IF, Western Blot
TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
Specifications
Specifications
| Antigen | TRA-1-60 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | TRA-1-60 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| Gene Alias | TRA160; tra60 |
| Host Species | Mouse |
| Immunogen | Human embryonal carcinoma cell line 2102Ep. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?