missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TR4/NR2C2 Antibody (OTI4B1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74577IR
This item is not returnable.
View return policy
Description
TR4/NR2C2 Monoclonal specifically detects TR4/NR2C2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TR4/NR2C2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NR2C2 | |
| Recombinant protein fragment corresponding to amino acids 1-241 of human TR4/NR2C2 (NP_003289) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI4B1 | |
| Western Blot, Immunohistochemistry | |
| hTAK1, Nuclear hormone receptor TR4, nuclear receptor subfamily 2 group C member 2, nuclear receptor subfamily 2, group C, member 2, Orphan nuclear receptor TAK1, Orphan nuclear receptor TR4, TAK1TR4TR2R1, Testicular receptor 4, TR4 nuclear hormone receptor | |
| Mouse | |
| Affinity Purified | |
| Neuroscience | |
| 7182.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction