missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527839
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (82%), Rat (81%), Chimpanzee (97%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a nuclear hormone receptor characterized by a highly conserved DNA binding domain (DBD), a variable hinge region, and a carboxy-terminal ligand binding domain (LBD) that is typical for all members of the steroid/thyroid hormone receptor superfamily. This protein also belongs to a large family of ligand-inducible transcription factors that regulate gene expression by binding to specific DNA sequences within promoters of target genes. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these variants has not been determined.
Specifications
| TR2 | |
| Polyclonal | |
| Unconjugated | |
| Nr2c1 | |
| 4831444H07Rik; 80.3; 80-3 cNDA; ATAR; CD270; CD40-like protein; early embryonic nuclear receptor; Eenr; herpes virus entry mediator A; Herpesvirus entry mediator A; HVEA; HVEM; LIGHTR; mTR2; nr2c1; Nuclear receptor subfamily 2 group C member 1; nuclear receptor subfamily 2, group C isoform; nuclear receptor subfamily 2, group C, member 1; nuclear receptor subfamily 2, group H, member 1; orphan nuclear receptor TR2; orphan receptor, TR2-11; pregnancy specific beta-1-glycoprotein 4; Psg4; testicular receptor 2; TNF receptor superfamily member 14; TNFRSF14; Tr2; TR2 nuclear hormone receptor; Tr2-11; Tumor necrosis factor receptor superfamily member 14; tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator); Tumor necrosis factor receptor-like 2; tumor necrosis factor receptor-like gene2; UNQ329/PRO509 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7181 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.8 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P13056 | |
| Nr2c1 | |
| Recombinant protein encompassing a sequence within the center region of human TR2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction