missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TR2/NR2C1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71804R
This item is not returnable.
View return policy
Description
TR2/NR2C1 Polyclonal antibody specifically detects TR2/NR2C1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TR2/NR2C1 | |
| Polyclonal | |
| Western Blot | |
| nuclear receptor subfamily 2 group C member 1, nuclear receptor subfamily 2, group C isoform, nuclear receptor subfamily 2, group C, member 1, Orphan nuclear receptor TR2, Testicular receptor 2, TR2, TR2 nuclear hormone receptor, TR2-11 | |
| The immunogen for this product maps to a region between residue 553 and 603 of human Testicular Receptor 2 using the numbering given in entry NP_003288.2 (GeneID 7181). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7181 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction