missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TR2/NR2C1 Polyclonal antibody specifically detects TR2/NR2C1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | TR2/NR2C1 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | nuclear receptor subfamily 2 group C member 1, nuclear receptor subfamily 2, group C isoform, nuclear receptor subfamily 2, group C, member 1, Orphan nuclear receptor TR2, Testicular receptor 2, TR2, TR2 nuclear hormone receptor, TR2-11 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 1 and 50 of human Testicular Receptor 2 using the numbering given in entry NP_003288.2 (GeneID 7181). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?