missing translation for 'onlineSavingsMsg'
Learn More

TR beta 1/NR1A2/Thyroid Hormone Receptor beta Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18354144 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354144 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354144 Supplier Novus Biologicals Supplier No. NBP310413100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TR beta 1/NR1A2/Thyroid Hormone Receptor beta Polyclonal specifically detects TR beta 1/NR1A2/Thyroid Hormone Receptor beta in Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TR beta 1/NR1A2/Thyroid Hormone Receptor beta
Applications ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation
Formulation PBS buffer, 2% sucrose
Gene Alias avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, c-erbA-2, c-erbA-beta, ERBA2MGC126109, ERBA-BETA, GRTH, MGC126110, NR1A2thyroid hormone receptor, beta (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog 2), Nuclear receptor subfamily 1 group A member 2, oncogene ERBA2, PRTH, THR1pituitary resistance to thyroid hormone, THRB1, THRB2, thyroid hormone receptor beta, thyroid hormone receptor beta 1, thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a)oncogene homolog 2, avian)
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the n terminal region of mouse TR beta 1/NR1A2/Thyroid Hormone Receptor beta (NP_033406). Peptide sequence MNYCMPEVHEVCPAASSNCYMQVTDYLAYLEDSPALSGRDVQAVPSSSIY
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7068
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.