missing translation for 'onlineSavingsMsg'
Learn More

TR alpha/NR1A1/Thyroid Hormone Receptor alpha Antibody, Novus Biologicals™

Product Code. 18408311 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
50 ug
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408311 50 ug 50µg
18127441 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18408311 Supplier Novus Biologicals Supplier No. NBP22252350ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TR alpha/NR1A1/Thyroid Hormone Receptor alpha Polyclonal specifically detects TR alpha/NR1A1/Thyroid Hormone Receptor alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TR alpha/NR1A1/Thyroid Hormone Receptor alpha
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:500
Formulation PBS with 1 mg/ml BSA. with 0.05% Sodium Azide
Gene Alias AR7, c-ERBA-1, c-erbA-alpha, EAR7, EAR-7, EAR-7.1/EAR-7.2, ERBA, ERBA1thyroid hormone receptor, alpha (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog), ERBA-related 7, MGC000261, MGC43240, NR1A 1, NR1A1THRA3, Nuclear receptor subfamily 1 group A member 1, THRA1ERB-T-1, THRA2, thyroid hormone receptor alpha, thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a)oncogene homolog, avian), triiodothyronine receptor, V-erbA-related protein 7
Gene Symbols THRA
Host Species Rabbit
Immunogen Synthetic Peptide: S(455) R A V C G E D D S S E A S S(469)
Purification Method Affinity Purified
Quantity 50 ug
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7067
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.