missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TR alpha/NR1A1/Thyroid Hormone Receptor alpha Antibody (H43), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79822AF594
This item is not returnable.
View return policy
Description
TR alpha/NR1A1/Thyroid Hormone Receptor alpha Monoclonal antibody specifically detects TR alpha/NR1A1/Thyroid Hormone Receptor alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| TR alpha/NR1A1/Thyroid Hormone Receptor alpha | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| H43 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| AR7, c-ERBA-1, c-erbA-alpha, EAR7, EAR-7, EAR-7.1/EAR-7.2, ERBA, ERBA1thyroid hormone receptor, alpha (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog), ERBA-related 7, MGC000261, MGC43240, NR1A 1, NR1A1THRA3, Nuclear receptor subfamily 1 group A member 1, THRA1ERB-T-1, THRA2, thyroid hormone receptor alpha, thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a)oncogene homolog, avian), triiodothyronine receptor, V-erbA-related protein 7 | |
| Synthetic peptide from the full length Human Thyroid hormone receptor protein | |
| 0.1 mL | |
| Apoptosis, Cancer, GPCR, Neuroscience | |
| 7067 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction