missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TQMN ADV MIRNA CARD FRMT 96A

This is a customizable panel for TaqMan Advanced miRNA assays, on a TAC card Visit our custom ordering page to reorder this or a similar product.

Brand:  Applied Biosystems™ A34724

Additional Details : Weight : 0.45360kg

Product Code. 15910453

  • £724.00 / Pack of 10
Estimated Shipment: 13-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This is a customizable panel for TaqMan Advanced miRNA assays, on a TAC card Visit our custom ordering page to reorder this or a similar product.

Specifications

Specifications

The TaqMan MicroRNA Arrays are shipped at ambient temperature. Upon arrival, store arrays at 2° to 8°C and keep them protected from light.
TaqMan™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers