Learn More
Applied Biosystems™ TQMN ADV MIRNA CARD FRMT 32
This is a customizable panel for TaqMan Advanced miRNA assays, on a TAC card Visit our custom ordering page to reorder this or a similar product.
Brand:  Applied Biosystems™ A34721
Additional Details : Weight : 0.45360kg
Product Code. 15980443
  • £694.00 / Pack of 10
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
This is a customizable panel for TaqMan Advanced miRNA assays, on a TAC card Visit our custom ordering page to reorder this or a similar product.
Specifications
Specifications
The TaqMan MicroRNA Arrays are shipped at ambient temperature. Upon arrival, store arrays at 2° to 8°C and keep them protected from light.
TaqMan™