missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TPX2 Recombinant Rabbit Monoclonal Antibody (0K5B6)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557999

 View more versions of this product

Product Code. 30284412

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MSQVKSSYSY DAPSDFINFS SLDDEGDTQN IDSWFEEKAN LENKLLGKNG TGGLFQGKTP LRKANLQQAI VTPLKPVDNT YYKEAEKENL VEQSIPSNAC SSLEVEAAIS RKTPAQPQRR SLRLSAQKDL EQKEKHHVKM KAKRCATPVI IDEILPSKKM KVSNNKKKPE EEGSAHQDTA EKNASSPEKA KGRHTVPCMP PAKQKFLKST EEQELEKSMK MQQEVVEMRK KNEEFKKLAL AGIGQPVKKS.

The protein encoded by this gene is found in the nucleolus and is similar to that of a family of genes on the Y-chromosome. This gene is intronless. Defects in this gene are a cause of sudden infant death with dysgenesis of the testes syndrome (SIDDT).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TPX2
Recombinant Monoclonal
1.06 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Q9ULW0
Tpx2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human TPX2 (NP_036244.2).
20 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
0K5B6
Unconjugated
Tpx2
2610005B21Rik; C20orf1; C20orf2; Differentially expressed in cancerous and non-cancerous lung cells 2; differentially expressed in lung cells; DIL2; DIL-2; FLS353; GD: HCA519; GD:C20orf1; HCA519; HCTP4; Hepatocellular carcinoma-associated antigen 519; hepatocellular carcinoma-associated antigen 90; p100; preferentially expressed in colorectal cancer; protein fls353; REPP86; restricted expression proliferation associated protein 100; restricted expression proliferation-associated protein 100; Targeting protein for Xklp2; TPX 2; Tpx2; TPX2 microtubule nucleation factor; TPX2, microtubule nucleation factor; TPX2, microtubule-associated; TPX2, microtubule-associated protein homolog; TPX2, microtubule-associated protein homolog (Xenopus laevis); TPX2, microtubule-associated, homolog
Rabbit
Protein A
RUO
22974
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.