missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPX2 Mouse anti-Human, Clone: 18D5, Abnova™
Mouse monoclonal antibody raised against partial recombinant human TPX2.
Brand: Abnova MAB12951.50ug
This item is not returnable.
View return policy
Specifications
| TPX2 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user. | |
| C20orf1/C20orf2/DIL-2/DIL2/FLS353/GD:C20orf1/HCA519/HCTP4/REPP86/p100 | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 22974 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 18D5 | |
| Mouse monoclonal antibody raised against partial recombinant human TPX2. | |
| In PBS, pH 7.2 (0.09% sodium azide). | |
| TPX2 | |
| Recombinant protein corresponding to amino acids 1-220 of human TPX2. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction