missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPTE2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25120-1-AP-150UL
This item is not returnable.
View return policy
Description
TPIP is a member of a large class of membrane-associated phosphatases with substrate specificity for the 3-position phosphate of inositol phospholipids.[supplied by OMIM, Jul 2002]Specifications
| TPTE2 | |
| Polyclonal | |
| Unconjugated | |
| TPTE2 | |
| Lipid phosphatase TPIP, TPIP, TPTE2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 93492 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6XPS3 | |
| TPTE2 | |
| TPTE2 Fusion Protein Ag19169 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction