missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPTE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566988
This item is not returnable.
View return policy
Description
Immunogen sequence: ERAYDPKHFHN RVSRIMIDDH NV Highest antigen sequence identity to the following orthologs - mouse 91%, rat 83%.
TPIP is a member of a large class of membrane-associated phosphatases with substrate specificity for the 3-position phosphate of inositol phospholipids.[supplied by OMIM, Jul 2002].
Specifications
| TPTE2 | |
| Polyclonal | |
| Unconjugated | |
| TPTE2 | |
| Lipid phosphatase TPIP; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase TPTE2; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase TPTE2; PTEN-like inositol lipid phosphatase; TPIP; TPIP lipid phosphatase; TPTE and PTEN homologous inositol lipid phosphatase; TPTE2; transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 93492 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6XPS3 | |
| TPTE2 | |
| Recombinant Human TPTE2. Recombinant protein control fragment (Product #RP-105652). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction