missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPTE2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | TPTE2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18231242
|
Novus Biologicals
NBP2-57829 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676748
|
Novus Biologicals
NBP2-57829-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TPTE2 Polyclonal specifically detects TPTE2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| TPTE2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 93492 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ERAYDPKHFHNRVSRIMIDDHNV | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase TPTE2, TPIP, transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 | |
| TPTE2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title