missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPTE2 Antibody [DyLight 350], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-45747UV
This item is not returnable.
View return policy
Description
TPTE2 Polyclonal antibody specifically detects TPTE2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| TPTE2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 93492 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot | |
| DyLight 350 | |
| phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase TPTE2, TPIP, transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 | |
| transmembrane phosphoinositide 3-phosphatase and tensin homolog 2. (Uniprot# Q6XPS3) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction