missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPST1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572357
This item is not returnable.
View return policy
Description
TPST1 Polyclonal Antibody for Western Blot
The tyrosylprotein sulfotransferases TPST-1 and TPST-2 catalyze the sulfation of tyrosine residues within secreted and membrane-bound proteins, such as cell adhesion molecules, G-protein-coupled receptors, coagulation factors, serpins, extracellular matrix proteins, and hormones. Although both TPST-1 and TPST-2 utilize 3'-phosphoadenosine 5'-phosphosulfate as their sulfate donor, they differ in their substrate specificity.
Specifications
| TPST1 | |
| Polyclonal | |
| Unconjugated | |
| TPST1 | |
| Protein-tyrosine sulfotransferase 1; R75054; TANGO13A; Tpst1; TPST-1; transport and golgi organization 13 homolog A; tyrosylprotein sulfotransferase 1; tyrosylprotein sulfotransferase-1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 22021, 8460 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O60507, O70281 | |
| TPST1 | |
| KLH conjugated synthetic peptide between 336-366 amino acids from the C-terminal region of human TPST1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction