Learn More
TPRKB Antibody (OTI3H3), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-74574JF549
Product Code. 18117755
  • £595.00 / 0.10mL
Description
TPRKB Monoclonal specifically detects TPRKB in Human, Mouse, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TPRKB
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
TPRKB
Full length recombinant protein of human TPRKB (NP_057142) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Monkey, Primate
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI3H3
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
CGI-121, PRPK (p53-related protein kinase)-binding protein, PRPK-binding protein, TP53RK binding protein, TP53RK-binding protein
Mouse
Affinity Purified
Signal Transduction
51002.0
Store at 4°C in the dark.