missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPRA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106836
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR175.
TPRA1 gene ontology (GO) annotations include aging; embryonic cleavage; lipid metabolic process; negative regulation of mitotic cell cycle phase transition.
Specifications
| TPRA1 | |
| Polyclonal | |
| Unconjugated | |
| TPRA1 | |
| 40kDa; FLJ32197; G protein-coupled receptor 175; GPR175; Integral membrane protein GPR175; PP6566; seven transmembrane domain orphan receptor; TMEM227; TPRA1; Tpra40; Transmembrane domain protein of 40 kDa regulated in adipocytes; transmembrane domain protein regulated in adipocytes 40 kDa; transmembrane protein 227; transmembrane protein adipocyte associated 1; transmembrane protein adipocyte-associated 1; transmembrane protein, adipocyte asscociated 1; transmembrane protein, adipocyte associated 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 131601, 24100, 85494 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q791F6, Q86W33, Q99MU1 | |
| TPRA1 | |
| A synthesized peptide derived from human TPRA1(Accession Q86W33), corresponding to amino acid residues F193-G243. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction