missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPRA1 Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Rabbit (100%) Dog, Bovine, Elephant, Panda, Horse (94%).
TPRA1 gene ontology (GO) annotations include aging; embryonic cleavage; lipid metabolic process; negative regulation of mitotic cell cycle phase transition.
Specifications
Specifications
| Antigen | TPRA1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | TPRA1 |
| Gene Accession No. | Q86W33 |
| Gene Alias | 40kDa; FLJ32197; G protein-coupled receptor 175; GPR175; Integral membrane protein GPR175; PP6566; seven transmembrane domain orphan receptor; TMEM227; TPRA1; Tpra40; Transmembrane domain protein of 40 kDa regulated in adipocytes; transmembrane domain protein regulated in adipocytes 40 kDa; transmembrane protein 227; transmembrane protein adipocyte associated 1; transmembrane protein adipocyte-associated 1; transmembrane protein, adipocyte asscociated 1; transmembrane protein, adipocyte associated 1 |
| Gene Symbols | TPRA1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?