missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-2866
This item is not returnable.
View return policy
Description
TPR Polyclonal specifically detects TPR in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockdown Validated.
Specifications
| TPR | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| nuclear pore complex-associated protein TPR, nucleoprotein TPR, translocated promoter region (to activated MET oncogene), tumor potentiating region | |
| Rabbit | |
| 267 kDa | |
| 0.1 mL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Panda, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, European domestic ferret, African elephant and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Knockout Validated | |
| NP_003283.1 | |
| TPR | |
| The immunogen recognized by this antibody maps to a region between residue 2275 and 2325 of human Translocated Promoter Region Protein using the numbering given in entry NP_003283.1 (GeneID 7175). | |
| Affinity Purified | |
| RUO | |
| 7175 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction