missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA561560
This item is not returnable.
View return policy
Description
Immunogen sequence: ATKRFKGKSK EEAFDAICQL VAGKEPANVG VTKAKTGGAV DRL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.
TPPP3 binds tubulin and has microtubule bundling activity. TPPP3 may play a role in cell proliferation and mitosis.
Specifications
| TPPP3 | |
| Polyclonal | |
| Unconjugated | |
| Tppp3 | |
| 2700055K07Rik; brain specific protein; Ceacam9; CGI-38; hypothetical protein LOC614988; mmCGM8; p20; p25gamma; RGD1305061; TPPP/p20; TPPP3; tubulin polymerization promoting protein family member 3; tubulin polymerization-promoting protein family member 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51673 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BW30 | |
| Tppp3 | |
| Recombinant protein corresponding to Human TPPP3 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction