missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPP3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£180.00 - £409.00
Specifications
| Antigen | TPPP3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228646
|
Novus Biologicals
NBP3-33391-20ul |
20 μL |
£180.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231068
|
Novus Biologicals
NBP3-33391-100ul |
100 μL |
£409.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TPPP3 Monoclonal antibody specifically detects TPPP3 in Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| TPPP3 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 51673 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| brain specific protein, CGI-38, p20, p25gamma, TPPP/p20, tubulin polymerization-promoting protein family member 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-176 of human TPPP3 (NP_057048.2).,, Sequence:, MAASTDMAGLEESFRKFAIHGDPKASGQEMNGKNWAKLCKDCKVADGKSVTGTDVDIVFSKVKGKSARVINYEEFKKALEELATKRFKGKSKEEAFDAICQLVAGKEPANVGVTKAKTGGAVDRLTDTSRYTGSHKERFDESGKGKGIAGRQDILDDSGYVSAYKNAGTYDAKVKK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title