missing translation for 'onlineSavingsMsg'
Learn More

TPPP3 Antibody, HRP, Novus Biologicals™

Product Code. 18333576 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333576 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333576 Supplier Novus Biologicals Supplier No. NBP300282H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TPPP3 Polyclonal specifically detects TPPP3 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPPP3
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias brain specific protein, CGI-38, p20, p25gamma, TPPP/p20, tubulin polymerization-promoting protein family member 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant TPPP3 (Q9BW30; Met1-Lys176).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51673
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.