missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPPP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557726
This item is not returnable.
View return policy
Description
Immunogen sequence: AKKRFKDKSS EEAVREVHRL IEGKAPIISG VTKAISSPTV SRLTDTTKFT GSHK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Tubulin family members are globular proteins important in the assembly of microtubules. Microtubules are structural components that play important roles in mitosis, cytokinesis and vesicle transport. TPPP (Tubulin polymerization-promoting protein), also known as p24 and p25, is a widely expressed 219 amino acid protein found in the perinuclear region of the cytoplasm. TPPP may form dimers and functions in polymerizing tubulin into double-walled tubules, polymorphic aggregates, or stabilized blocks. TPPP overexpression prevents formation of the mitotic spindle assembly and breakdown of the nuclear envelope. TPPP is phosphorylated by TPK II and is encoded by a gene that maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. May play a role in the polymerization of tubulin into microtubules, microtubule bundling and the stabilization of existing microtubules, thus maintaining the integrity of the microtubule network. May play a role in mitotic spindle assembly and nuclear envelope breakdown.
Specifications
| TPPP | |
| Polyclonal | |
| Unconjugated | |
| TPPP | |
| 25 kDa brain-specific protein; 2900041A09Rik; AI849835; brain specific protein p25 alpha; brain-specific protein p25 alpha; glycogen synthase kinase 3 (GSK3) inhibitor p24; p24; p25; p25alpha; p25-alpha; RGD1310121; Tppp; TPPP/p25; TPPP1; tubulin polymerization promoting protein; tubulin polymerization-promoting protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11076 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94811 | |
| TPPP | |
| Recombinant protein corresponding to Human TPPP. Recombinant protein control fragment (Product #RP-96956). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction