missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPP2 Recombinant Rabbit Monoclonal Antibody (24GB2015)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557798
This item is not returnable.
View return policy
Description
TPP2 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.
Specifications
| TPP2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TPP2 | |
| TPP2; TPP-2; TppII; TPP-II; Tripeptidyl aminopeptidase; tripeptidyl peptidase 2; tripeptidyl peptidase II; tripeptidyl-peptidase 2; Tripeptidyl-peptidase II | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22019, 7174, 81815 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2015 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P29144, Q64514, Q64560 | |
| TPP2 | |
| A synthesized peptide derived from human Tripeptidyl peptidase II (aa 900-1150). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction