missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPP1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00001200-B01P.50ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MGLQACLLGLFALILSGKCSYSPEPDQRRTLPPGWVSLGRADPEEELSLTFALRQQNVERLSELVQAVSDPSSPQYGKYLTLENVADLVRPSPLTLHTVQKWLLAAGAQKCHSVITQDFLTCWLSIRQAELLLPGAEFHHYVGGPTETHVVRSPHPYQLPQALAPHVDFVGGLHRFPPTSSLRQRPEPQVTGTVGLHLGVTPSVIRKRYNLTSQDVGSGTSNNSQACAQFLEQYFHDSDLAQFMRLFGGNFAHQASVARVVGQQGRGRAGIEASLDVQYLMSAGANISTWVYSSPGRHEGQEPFLQWLMLLSNESALPHVHTVSYGDDEDSLSSAYIQRVNTELMKAAARGLTLLFASGDSGAGCWSVSGRHQFRPTFPASSPYVTTVGGTSFQEPFLITNEIVDYISGGGFSNVFPRPSYQEEAVTKFLSSSPHLPPSSYFNASGRAYPDVAALSDGYWVVSNRVPIPWVSGTSASTPVFGGILSLINEHRILSGRPPLGFLNPRLYQQHGAGLFDVTRGCHESCLDEEVEGQGFCSGPGWDPVTGWGTPNFPALLKTLLNPSpecifications
TPP1 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
NM_000391 | |
TPP1 | |
TPP1 (NP_000382.3, 1 a.a. ∼ 563 a.a) full-length human protein. | |
RUO | |
1200 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
tripeptidyl peptidase I | |
CLN2/GIG1/LPIC/MGC21297 | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |