Learn More
TPMT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA587693
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MDGTRTSLDI EEYSDTEVQK NQVLTLEEWQ DKWVNGKTAF HQEQGHQLLK KHLDTFLKGK SGLRVFFPLC GKAVEMKWFA DRGHSVVGVE ISELGIQEFF TEQNLSYSEE PITEIPGTKV FKSSSGNISL YCCSIFDLPR TNIGKFDMIW DRGALVAINP GDRKCYADTM FSLLGKKFQY LLCVLSYDPT KHPGPPFYVP HAEIERLFGK ICNIRCLEKV DAFEERHKSW GIDCLFEKLY LLTEK; Positive Samples: K562; Cellular Location: Cytoplasm
Catalyzes the S-methylation of thiopurine drugs such as 6-mercaptopurine. Defects in TPMT are the cause of thiopurine S-methyltransferase deficiency (TPMT deficiency). TPMT is an enzyme involved in the normal metabolic inactivation of thiopurine drugs. These drugs are generally used as immunosupressants or cytotoxic drugs and are prescribed for a variety of clinical conditions including leukemia, autoimmune disease and organ transplantation. Patients with intermediate or no TPMT activity are at risk of toxicity after receiving standard doses of thiopurine drugs and it is shown that inter-individual differences in response to these drugs are largely determined by genetic variation at the TPMT locus.Specifications
TPMT | |
Polyclonal | |
Unconjugated | |
TPMT | |
Tpmt | |
Recombit fusion protein containing a sequence corresponding to amino acids 1-245 of human TPMT | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Western Blot | |
4.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O55060, P51580 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22017, 7172 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |