missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPMT Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68251-1-IG-20UL
This item is not returnable.
View return policy
Description
The TPMT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TPMT. This antibody recognizes human antigen. The TPMT antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| TPMT | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 7172 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1D4B3 | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 32 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction