missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TPMT Polyclonal antibody specifically detects TPMT in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antigen | TPMT |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:10 - 1:100 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-245 of human TPMT (NP_000358.1).,, Sequence:, MDGTRTSLDIEEYSDTEVQKNQVLTLEEWQDKWVNGKTAFHQEQGHQLLKKHLDTFLKGKSGLRVFFPLCGKAVEMKWFADRGHSVVGVEISELGIQEFFTEQNLSYSEEPITEIPGTKVFKSSSGNISLYCCSIFDLPRTNIGKFDMIWDRGALVAINPGDRKCYADTMFSLLGKKFQYLLCVLSYDPTKHPGPPFYVPHAEIERLFGKICNIRCLEKVDAFEERHKSWGIDCLFEKLYLLTEK |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?