missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TPMT Antibody (2H3), Novus Biologicals™

Product Code. 18345088 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345088 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345088 Supplier Novus Biologicals™ Supplier No. H00007172M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TPMT Monoclonal antibody specifically detects TPMT in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPMT
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 2H3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05339
Gene Alias EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase
Host Species Mouse
Immunogen TPMT (AAH05339, 1 a.a. ∽ 245 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDGTRTSLDIEEYSDTEVQKNQVLTLEEWQDKWVNGKTAFHQEQGHQLLKKHLDTFLKGKSGLRVFFPLCGKAVEMKWFADRGHSVVGVEISELGIQEFFTEQNLSYSEEPITEIPGTKVFKSSSGNISLYCCSIFDLPRTNIGKFDMIWDRGALVAINPGDRKCYADTMFSLLGKKFQYLLCVLSYDPTKHPGPPFYVPHAEIERLFGKICNIRRLEKVDAFEERHKSWGIDCLFEKLYLLTEK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7172
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.