missing translation for 'onlineSavingsMsg'
Learn More

TPMT Antibody (2E7), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-97939AF405

Product Code. 18026907

  • £418.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TPMT Monoclonal specifically detects TPMT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TPMT
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
TPMT
Recombinant human TPMT (1-245aa) purified from E. coli
100 μL
Proteases & Other Enzymes, Signal Transduction
7172
Human
Purified
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
20000000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase
Mouse
Protein G purified
RUO
Primary
Anti-human TPMT mAb, clone 2E7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TPMT protein.
Store at 4C in the dark.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.