missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TPMPA
GREENER_CHOICE

Product Code. 30001170
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001170

Brand: Tocris Bioscience™ 1040/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective GABAA-ρ antagonist

A selective, competitive GABAA-ρ antagonist with only minimal effects on GABAA and GABAB receptors (Kb values are 2.1 μM (antagonist), 320 μM (antagonist) and EC50 ∽ 500 μM (weak agonist) respectively). Displays 8-fold selectivity for human recombinant ρ1 receptors over ρ2 receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182485-36-5
Molecular Formula C6H12NO2P

Specifications

Quantity 10 mg
Formula Weight 161.14
Purity >98%
Molecular Weight (g/mol) 161.141
Chemical Name or Material TPMPA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.