Learn More
TPM4 Mouse anti-Human, Unconjugated, Clone: 2E12D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67244-1-IG-20UL
This item is not returnable.
View return policy
Description
The TPM4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TPM4. This antibody recognizes human, mouse, rat, pig antigen. The TPM4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TPM4 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, ELISA | |
| 2E12D12 | |
| Unconjugated | |
| 20 μL | |
| 7171 | |
| IgG1 |