missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA591925
This item is not returnable.
View return policy
Description
Positive Samples: SW620, HeLa, MCF7, U-87MG, Mouse skeletal muscle, Mouse lung; Cellular Location: Cytoplasm, cytoskeleton Immunogen sequence: MDAIKKKMQM LKLDKENAID RAEQAEADKK QAEDRCKQLE EEQQALQKKL KGTEDEVEKY SESVKEAQEK LEQAEKKATD AEADVASLNR RIQLVEEELD.
TPM2 (tropomyosin 2) is beta-tropomyosin, an isoform of tropomyosin that is mainly expressed in slow, type 1 muscle fibers. TPM2 binds to actin filaments in muscle and non-muscle cells. Plays a central role in association with the troponin complex in the calcium dependent regulation of vertebrate striated muscle contraction. Smooth muscle contraction is regulated by interaction with caldesmon. Mutations in this gene can cause nemaline myopathy 4, arthrogryposis dital 1A/2B, and cap myopathy 2.
Specifications
| TPM2 | |
| Polyclonal | |
| Unconjugated | |
| TPM2 | |
| AMCD1; beta tropomyosin; beta-tropomyosin; cb836; cytoskeletal protein; DA1; DA2B; epididymis secretory protein Li 273; fj63f03; HEL-S-273; NEM4; nemaline myopathy type 4; RP11-112J3.4; TMSB; TPM2; Tpm-2; tpm4l; Trop-2; tropomyosin; tropomyosin 2; tropomyosin 2 (beta); tropomyosin 2, beta; tropomyosin beta chain; Tropomyosin-2; unnamed protein product; wu:fj63f03; zgc:86810 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22004, 500450, 7169 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P07951, P58774, P58775 | |
| TPM2 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction