missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55443-1-AP-20UL
This item is not returnable.
View return policy
Description
TPM2 (tropomyosin 2) is beta-tropomyosin, an isoform of tropomyosin that is mainly expressed in slow, type 1 muscle fibers. TPM2 binds to actin filaments in muscle and non-muscle cells. Plays a central role in association with the troponin complex in the calcium dependent regulation of vertebrate striated muscle contraction. Smooth muscle contraction is regulated by interaction with caldesmon. Mutations in this gene can cause nemaline myopathy 4, arthrogryposis dital 1A/2B, and cap myopathy 2.Specifications
| TPM2 | |
| Polyclonal | |
| Unconjugated | |
| TPM2 | |
| AMCD1, Beta tropomyosin, DA1, DA2B, TMSB, TPM2, Tropomyosin 2, tropomyosin 2 (beta), Tropomyosin beta chain | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22004, 7169 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07951, P58774 | |
| TPM2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction