missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115541
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MAP3K8/COT.
MAP3K8 (COT) is a serine/threonine kinase which can activate both the MAP and JNK kinase pathways and plays a role in the cell cycle. Mitogen-activated protein kinase (MAPK) signaling cascades include MAPK or extracellular signal-regulated kinase (ERK), MAPK kinase (MKK or MEK), and MAPK kinase (MAPKKK or MEKK). MAPKK kinase/MEKK phosphorylates and activates its downstream protein kinase, MAPK kinase/MEK, which in turn activates MAPK. The kinases of these signaling cascades are highly conserved, and homologs exist in yeast, Drosophila, and mammalian cells. MEKK8 is able to activate NF-kappa-B 1 by stimulating proteasome-mediated proteolysis of NF-kappa-B 1/p105. This cytoplasmic protein is expressed in several normal tissues and human tumor-derived cell lines. The 58 kDa form is activated specifically during the S and G2/M phases of the cell cycle. The longer form undergoes phosphorylation on Ser residues mainly, and the shorter form on both Ser and Thr residues.
Specifications
| TPL2 | |
| Polyclonal | |
| Unconjugated | |
| MAP3K8 | |
| augmented in rheumatoid arthritis 2; AURA2; Cancer Osaka thyroid oncogene; cancer Osaka thyroid, oncogene; CCOT; c-COT; COT; cot (cancer Osaka thyroid) oncogene; COT proto-oncogene serine/threonine-protein kinase; Cot/Tpl2; D17TPL2; EC 1.14.16.2; Est; Estf; Ewing sarcoma transformant; FLJ10486; M3K8; MAP3K 8; Map3k8; MEKK8; mitogen activated protein kinase kinase kinase 8; mitogen-activated protein kinase kinase kinase 8; Protein kinase cot; Proto-oncogene c-Cot; proto-oncogene serine/threoine protein kinase; serine/threonine kinase (Tpl-2); serine/threonine-protein kinase cot; TH isoform 3; TH isoform a; TH-4; TPL 2; Tpl2; TPL-2; Tumor progression locus 2; TY3H; TYH | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116596, 1326, 26410 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P41279, Q07174, Q63562 | |
| MAP3K8 | |
| A synthesized peptide derived from human MAP3K8(Accession P41279), corresponding to amino acid residues T264-G314. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction