missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584039
This item is not returnable.
View return policy
Description
Immunogen sequence: GEEAEFHFAA LYISGQWPRL RADTDLQRLG SSAMAPSRKF FVGGNWKMNG RKQSLGELIG TLNAAKVPAD TEVVCAPPTA YIDFARQKLD P Highest antigen sequence indentity to the following orthologs: Rat - 78%, Mouse - 77%.
This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.
Specifications
| TPI1 | |
| Polyclonal | |
| Unconjugated | |
| Tpi1 | |
| AI255506; epididymis secretory protein Li 49; HEL-S-49; Methylglyoxal synthase; MGC88108; TIM; Tpi; TPI1; Tpi-1; TPID; triosephosphate isomerase; triose-phosphate isomerase; triosephosphate isomerase 1; triose-phosphate isomerase TPI1; unnamed protein product; YD9609.05C; YDR050C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7167 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P60174 | |
| Tpi1 | |
| Recombinant protein corresponding to Human TPI1. Recombinant protein control fragment (Product #RP-104914). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction