missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526185
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Tryptophan hydroxylase (TPH) is a member of the pterin-dependent aromatic acid hydroxylase family. It catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. TPH is expressed predominantly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.
Specifications
| TPH2 | |
| Polyclonal | |
| Unconjugated | |
| Tph2 | |
| ADHD7; AU043594; MGC119994; Neuronal tryptophan hydroxylase; NTPH; Tph2; TPRH; TRPH; Tryptophan 5-hydroxylase 2; tryptophan 5-monooxygenase 2; tryptophan hydroxylase 2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 121278, 216343 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8CGV2, Q8IWU9 | |
| Tph2 | |
| KLH conjugated synthetic peptide between 168-193 amino acids from the central region of human TPH2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction