missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TPH2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17845952
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17845952 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17845952 Supplier Invitrogen™ Supplier No. PA5142778

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

Tryptophan hydroxylase (TPH) is a member of the pterin-dependent aromatic acid hydroxylase family. It catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. TPH is expressed predominantly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPH2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Tph2
Gene Accession No. Q8CGV2
Gene Alias ADHD7; AU043594; MGC119994; Neuronal tryptophan hydroxylase; NTPH; Tph2; TPRH; TRPH; Tryptophan 5-hydroxylase 2; tryptophan 5-monooxygenase 2; tryptophan hydroxylase 2
Gene Symbols Tph2
Host Species Goat
Immunogen Peptide with sequence C-SLTQNKAIKSEDK (aa 33-45).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216343
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.