missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPGS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566420
This item is not returnable.
View return policy
Description
Immunogen sequence: TQLTQSSMYSL PNAPTLADLE DDTHEASDDQ PEKPHFDSRS VIFELDSCNG SGKVCLVYKS GKPALAEDTE IWFLDRALYW HFL Highest antigen sequence identity to the following orthologs - mouse 84%, rat 81%.
This gene encodes a protein that is a component of the neuronal polyglutamylase complex, which plays a role in post-translational addition of glutamate residues to C-terminal tubulin tails. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| TPGS2 | |
| Polyclonal | |
| Unconjugated | |
| TPGS2 | |
| C18orf10; HMFN0601; HsT3006; L17; PGs2; TPGS2; Tubulin polyglutamylase complex subunit 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25941 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68CL5 | |
| TPGS2 | |
| Recombinant Human TPGS2. Recombinant protein control fragment (Product #RP-105206). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction