missing translation for 'onlineSavingsMsg'
Learn More

TPEN, MedChemExpress

Product Code. 16479496
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479496 50 mg 50mg
16424256 100 mg 100mg
16420906 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16479496 Supplier Medchem Express Supplier No. HY100202
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TPEN (TPEDA) is a specific cell-permeable heavy metal chelator. TPEN has a higher affinity for Zn2+, but a lower affinity for Mg2+ and Ca2+. TPEN induces DNA damage and increases intracellular ROS production. TPEN also inhibits cell proliferation and induces apoptosis.

TPEN (TPEDA) is a specific cell-permeable heavy metal chelator. TPEN has a higher affinity for Zn2+, but a lower affinity for Mg2+ and Ca2+. TPEN induces DNA damage and increases intracellular ROS production. TPEN also inhibits cell proliferation and induces apoptosis.

Specifications

Chemical Name or Material TPEN
Physical Form Solid
CAS 16858-02-9
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C26H28N6
For Use With (Application) Cancer-programmed cell death
Synonym TPEDA
Solubility Information DMSO : 20 mg/mL (47.11 mM; Need ultrasonic)
SMILES N(CC1=NC=CC=C1)(CC2=NC=CC=C2)CCN(CC3=NC=CC=C3)CC4=NC=CC=C4
Molecular Weight (g/mol) 424.54
Formula Weight 424.54
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.