missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPD52L3 Monoclonal Antibody (OTI8C12)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526024
This item is not returnable.
View return policy
Description
TPD52L3 Monoclonal Antibody for Western Blot, IHC (P), Flow
Belongs to the TPD52 family and is specifically expressed in testis. Expressed at 5.6-fold higher levels in adult testis than in fetal testis.
Specifications
| TPD52L3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96J77 | |
| TPD52L3 | |
| Full length human recombinant protein of TPD52L3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI8C12 | |
| Unconjugated | |
| TPD52L3 | |
| d55; hD55; NYDSP25; protein kinase NYD-SP25; RGD1309391; Testis development protein NYD-SP25; testis tissue sperm-binding protein Li 87P; Tpd52l3; Trpd52l3; Tumor protein D52-like 3; tumor protein D55 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 89882 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction