missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPD52L3/D55 Antibody (OTI8C12), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74567DL594
This item is not returnable.
View return policy
Description
TPD52L3/D55 Monoclonal antibody specifically detects TPD52L3/D55 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications
| TPD52L3/D55 | |
| Monoclonal | |
| DyLight 594 | |
| hD55, MGC26757, NYDSP25, NYD-SP25, protein kinase NYD-SP25, Testis development protein NYD-SP25, tumor protein D52-like 3MGC45374, tumor protein D55 | |
| Full length recombinant protein of human TPD52L3/D55 (NP_001001875) produced in HEK293T cells. | |
| 0.1 mL | |
| Protein Kinase | |
| 89882 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| OTI8C12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction