missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPD52L3/D55 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35573-100ul
This item is not returnable.
View return policy
Description
TPD52L3/D55 Polyclonal antibody specifically detects TPD52L3/D55 in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| TPD52L3/D55 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| hD55, MGC26757, NYDSP25, NYD-SP25, protein kinase NYD-SP25, Testis development protein NYD-SP25, tumor protein D52-like 3MGC45374, tumor protein D55 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-132 of human TPD52L3/D55 (NP_001001875.1).,, Sequence:, MPHARTETSVGTYESHSTSELEDLTEPEQRELKTKLTKLEAEIVTLRHVLAAKERRCGELKRKLGLTALVGLRQNLSKSWLDVQVSNTYVKQKTSAALSTMGTLICRKLGGVKKSATLRSFEGNPKGEGSRI | |
| 100 μL | |
| Protein Kinase | |
| 89882 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction