missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPD52L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599301
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TPD54.
EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).
Specifications
| TPD52L2 | |
| Polyclonal | |
| Unconjugated | |
| Tpd52l2 | |
| 2810411G23Rik; AU016537; C86069; D54; DKFZp686A1765; HCCR-binding protein 2; hD54; Tpd52l2; TPD54; tumor protein D52 like 2; tumor protein D52-like 2; Tumor protein D54 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 296480, 66314, 7165 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43399, Q6PCT3, Q9CYZ2 | |
| Tpd52l2 | |
| A synthesized peptide derived from human TPD52L2(Accession O43399), corresponding to amino acid residues K175-F206. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction